ENA429359646 Enamine

3-(aminomethyl)-2-methyloxolan-3-ol

  • MDL number MFCD24293720
  • Cas Number : 1548849-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188324
Availability details for
Quantity
  • Availability
  • Add to Cart