ENA429359652 Enamine

3-chlorocyclohex-2-en-1-one

  • MDL number MFCD00052128
  • Cas Number : 5682-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187177
Availability details for
Quantity
  • Availability
  • Add to Cart