ENA429359655 Enamine

N-[(2-methoxyphenyl)methyl]-2-phenylcyclopropan-1-amine

  • MDL number MFCD16246356
  • Cas Number : 1306969-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188338

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188338
Availability details for
Quantity
  • Availability
  • Add to Cart