ENA429359661 Enamine

5-(ethylamino)pyrazine-2-carbonitrile

  • MDL number MFCD20283490
  • Cas Number : 1409987-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188346

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188346
Availability details for
Quantity
  • Availability
  • Add to Cart