ENA429359667 Enamine

methyl 3-sulfanylnaphthalene-2-carboxylate

  • MDL number MFCD26024175
  • Cas Number : 86163-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188352
Availability details for
Quantity
  • Availability
  • Add to Cart