ENA429359672 Enamine

5-(2,3-dihydro-1H-inden-5-yl)-1,2,4-oxadiazol-3-amine

  • MDL number MFCD18867092
  • Cas Number : 1340362-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188359
Availability details for
Quantity
  • Availability
  • Add to Cart