ENA429359673 Enamine

4-cyclobutoxybenzene-1-carbothioamide

  • MDL number MFCD16684499
  • Cas Number : 1247109-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188360

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188360
Availability details for
Quantity
  • Availability
  • Add to Cart