ENA429359674 Enamine

3-(3-chloro-4-fluorophenyl)-4,5-dihydro-1,2,4-oxadiazol-5-one

  • MDL number MFCD17291175
  • Cas Number : 1343830-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188361
Availability details for
Quantity
  • Availability
  • Add to Cart