ENA429359675 Enamine

3,3,3-trifluoro-2-hydroxy-2-methylpropanenitrile

  • MDL number MFCD00040885
  • Cas Number : 335-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188363

Safety Information

signalword Danger
safety_statement 36/37
msds https://www.enaminestore.com/catalog/EN300-188363
pictograms GHS02,GHS06,GHS09
Availability details for
Quantity
  • Availability
  • Add to Cart