ENA429359676 Enamine

4-amino-2-(propan-2-yloxy)benzamide

  • MDL number MFCD19640755
  • Cas Number : 1319746-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188364

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188364
Availability details for
Quantity
  • Availability
  • Add to Cart