ENA429359677 Enamine

3-(trifluoromethyl)-[1,2,4]triazolo[4,3-b]pyridazin-6-amine

  • MDL number MFCD03001630
  • Cas Number : 889943-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188366
Availability details for
Quantity
  • Availability
  • Add to Cart