ENA429359681 Enamine

3-(4-chlorophenyl)-8-azabicyclo[3.2.1]octan-3-ol hydrochloride

  • MDL number MFCD28118717
  • Cas Number : 1795278-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188370
Availability details for
Quantity
  • Availability
  • Add to Cart