ENA429359683 Enamine

4-aminobut-2-enoic acid hydrochloride

  • MDL number MFCD12923255
  • Cas Number : 129564-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188373

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188373
Availability details for
Quantity
  • Availability
  • Add to Cart