ENA429359684 Enamine

3,5-dibromo-2-methoxythiophene

  • MDL number MFCD26127511
  • Cas Number : 77133-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188374
Availability details for
Quantity
  • Availability
  • Add to Cart