ENA429359686 Enamine

3-(5-bromothiophen-2-yl)-1-methyl-1H-pyrazol-5-amine

  • MDL number MFCD11209695
  • Cas Number : 1152666-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188376
Availability details for
Quantity
  • Availability
  • Add to Cart