ENA429359689 Enamine

methyl 6-hydroxy-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD16615464
  • Cas Number : 380419-63-6
  • Molecular Weight : 277.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-188382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188382
Availability details for
Quantity
  • Availability
  • Add to Cart