ENA429359690 Enamine

3-(3-iodo-4-methyl-1H-pyrazol-1-yl)-1$l^{6}-thiolane-1,1-dione

  • MDL number MFCD23769073
  • Cas Number : 1529378-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188384

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188384
Availability details for
Quantity
  • Availability
  • Add to Cart