ENA429359691 Enamine

but-3-yne-1-sulfonamide

  • MDL number MFCD19613775
  • Cas Number : 1341659-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188385

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188385
Availability details for
Quantity
  • Availability
  • Add to Cart