ENA429359692 Enamine

but-2-yne-1-sulfonamide

  • MDL number MFCD19613773
  • Cas Number : 1341348-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188386

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188386
Availability details for
Quantity
  • Availability
  • Add to Cart