ENA429359696 Enamine

3-amino-N-methyl-3-phenylpropanamide

  • MDL number MFCD09945160
  • Cas Number : 180264-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188390
Availability details for
Quantity
  • Availability
  • Add to Cart