ENA429359697 Enamine

2-amino-3-methoxybenzonitrile

  • MDL number MFCD09744022
  • Cas Number : 148932-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-67
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188391

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188391
Availability details for
Quantity
  • Availability
  • Add to Cart