ENA429359713 Enamine

2,2-dimethylcyclohexane-1-carboxylic acid

  • MDL number MFCD20639457
  • Cas Number : 62581-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188485

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188485
Availability details for
Quantity
  • Availability
  • Add to Cart