ENA429359714 Enamine

1-{imidazo[1,2-a]pyrimidin-2-yl}ethan-1-one

  • MDL number MFCD23904666
  • Cas Number : 1521986-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188489
Availability details for
Quantity
  • Availability
  • Add to Cart