ENA429359717 Enamine

3-ethoxypropanal

  • MDL number MFCD01696143
  • Cas Number : 2806-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-188492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188492
Availability details for
Quantity
  • Availability
  • Add to Cart