ENA429359720 Enamine

4,5-dimethoxy-2-methylbenzene-1-sulfonamide

  • MDL number MFCD11647571
  • Cas Number : 60987-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188495

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188495
Availability details for
Quantity
  • Availability
  • Add to Cart