ENA429359721 Enamine

N-[(2-aminophenyl)methyl]-N-methyl-1,3-benzothiazol-2-amine

  • MDL number MFCD13158851
  • Cas Number : 1182913-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188496
Availability details for
Quantity
  • Availability
  • Add to Cart