ENA429359722 Enamine

3-[(2-amino-5-fluorophenyl)methyl]-2,3-dihydro-1,3-thiazol-2-one

  • MDL number MFCD18328103
  • Cas Number : 1338984-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188497
Availability details for
Quantity
  • Availability
  • Add to Cart