ENA429359726 Enamine

3-(aminomethyl)-1-ethyl-1,2-dihydropyridin-2-one hydrochloride

  • MDL number MFCD28118724
  • Cas Number : 1795440-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188501
Availability details for
Quantity
  • Availability
  • Add to Cart