ENA429359727 Enamine

[2-(aminomethyl)spiro[3.3]heptan-2-yl]methanol

  • MDL number MFCD21122125
  • Cas Number : 1481497-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188502

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188502
Availability details for
Quantity
  • Availability
  • Add to Cart