ENA429359728 Enamine

1-(3-fluoro-5-methylphenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD21784952
  • Cas Number : 1511171-91-7
  • Molecular Weight : 221.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-188503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188503
Availability details for
Quantity
  • Availability
  • Add to Cart