ENA429359729 Enamine

2,5,7-trichloro-1,3-benzoxazole

  • MDL number MFCD11890752

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188504
Availability details for
Quantity
  • Availability
  • Add to Cart