ENA429359732 Enamine

2,2-diethoxyethanimidamide hydrochloride

  • MDL number MFCD10697950
  • Cas Number : 203054-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188507

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-188507
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart