ENA429359733 Enamine

methyl 1-(bromomethyl)cyclopropane-1-carboxylate

  • MDL number MFCD16036400
  • Cas Number : 43161-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188508

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-188508
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart