ENA429359734 Enamine

2-{[2-(2-hydroxyethoxy)ethyl]sulfanyl}ethan-1-ol

  • MDL number MFCD12785326
  • Cas Number : 85391-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188510
Availability details for
Quantity
  • Availability
  • Add to Cart