ENA429359738 Enamine

3-(2,6-dimethylmorpholin-4-yl)-4,4,4-trifluorobutan-2-amine

  • MDL number MFCD16069025
  • Cas Number : 1249307-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188520
Availability details for
Quantity
  • Availability
  • Add to Cart