ENA429359739 Enamine

1H-pyrrolo[3,2-b]pyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD23135791
  • Cas Number : 860363-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188521

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188521
Availability details for
Quantity
  • Availability
  • Add to Cart