ENA429359740 Enamine

3-[4-formyl-3-(pyridin-4-yl)-1H-pyrazol-1-yl]propanenitrile

  • MDL number MFCD28118726
  • Cas Number : 1803566-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188527
Availability details for
Quantity
  • Availability
  • Add to Cart