ENA429359741 Enamine

2-(2,2-difluoroethyl)-2H,3H,5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]piperazin-3-one

  • MDL number MFCD21739756
  • Cas Number : 1498956-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188531

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188531
Availability details for
Quantity
  • Availability
  • Add to Cart