ENA429359742 Enamine

1-bromo-4-(difluoromethoxy)-2-fluorobenzene

  • MDL number MFCD18390897
  • Cas Number : 1242258-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188533

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-188533
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart