ENA429359743 Enamine

2-[4-(difluoromethoxy)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD22419269
  • Cas Number : 1628442-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188534

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188534
Availability details for
Quantity
  • Availability
  • Add to Cart