ENA429359753 Enamine

1-(1-benzothiophen-7-yl)ethan-1-one

  • MDL number MFCD18451765
  • Cas Number : 22720-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187178
Availability details for
Quantity
  • Availability
  • Add to Cart