ENA429359762 Enamine

1-(3-fluorophenyl)-1H-1,2,3-triazole-5-carboxylic acid

  • MDL number MFCD12116531
  • Cas Number : 1153490-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188597

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188597
Availability details for
Quantity
  • Availability
  • Add to Cart