ENA429359764 Enamine

N-[1-(aminomethyl)cyclopentyl]-3-methyl-[1,2,4]triazolo[4,3-a]pyrazin-8-amine dihydrochloride

  • MDL number MFCD28118736
  • Cas Number : 1803587-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188601
Availability details for
Quantity
  • Availability
  • Add to Cart