ENA429359766 Enamine

7-chlorothieno[2,3-c]pyridine

  • MDL number MFCD04973300
  • Cas Number : 28948-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188604

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-188604
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart