ENA429359769 Enamine

tert-butyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-1-carboxylate

  • MDL number MFCD24468572
  • Cas Number : 402848-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188608
Availability details for
Quantity
  • Availability
  • Add to Cart