ENA429359771 Enamine

tert-butyl 4-{[(6-chloropyridin-3-yl)oxy]methyl}piperidine-1-carboxylate

  • MDL number MFCD24450233
  • Cas Number : 1010115-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188610

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188610
Availability details for
Quantity
  • Availability
  • Add to Cart