ENA429359773 Enamine

1,2-dimethyl-1H-1,3-benzodiazole-6-carboxylic acid

  • MDL number MFCD13562914
  • Cas Number : 6595-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188615

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188615
Availability details for
Quantity
  • Availability
  • Add to Cart