ENA429359774 Enamine

1-(2,4-dimethylpyrimidin-5-yl)ethan-1-ol

  • MDL number MFCD17219516
  • Cas Number : 1310684-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188617

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188617
Availability details for
Quantity
  • Availability
  • Add to Cart