ENA429359775 Enamine

5-methyl-5H,6H,7H-furo[2,3-f]indol-6-one

  • MDL number MFCD07777108
  • Cas Number : 851682-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188618
Availability details for
Quantity
  • Availability
  • Add to Cart