ENA429359781 Enamine

(4-chloro-3-methoxyphenyl)hydrazine

  • MDL number MFCD19201855
  • Cas Number : 178423-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-188647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188647
Availability details for
Quantity
  • Availability
  • Add to Cart